Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cids-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128999.4 | NP_001122471.1 | 2 - 3 | 228 | 149 | |
NM_001129000.5 | NP_001122472.1 | 2 - 3 | 276 | 149 | ||
NM_059979.6 | NP_492380.2 | 2 - 3 | 237 | 149 | ||
NM_182083.6 | NP_871883.2 | 2 - 3 | 228 | 149 | ||
NR_023944.1 | 2 - 3 | 164 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |