Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059980.6 | NP_492381.3 | 12 - 13 | 1735 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |