Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059992.4 | NP_492393.1 | 12 - 13 | 2064 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |