Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059992.4 | NP_492393.1 | 16 - 17 | 2590 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |