Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tax-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060026.6 | NP_492427.3 | 12 - 13 | 2082 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |