Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026506.5 | NP_001021677.3 | 12 - 13 | 1991 | 88 | |
NM_060041.6 | NP_492442.3 | 12 - 13 | 2025 | 88 | ||
NM_060042.5 | NP_492443.3 | 12 - 13 | 2011 | 88 | ||
NM_060043.5 | NP_492444.4 | 13 - 14 | 2328 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Caenorhabditis elegans Probe spans exons |