Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026507.5 | NP_001021678.3 | 13 - 14 | 2435 | 88 | |
NM_001026508.6 | NP_001021679.3 | 12 - 13 | 1980 | 88 | ||
NR_000051.1 | 13 - 14 | 2215 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |