Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mel-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276534.2 | NP_001263463.1 | 2 - 3 | 543 | 92 | |
NM_060048.11 | NP_492449.2 | 3 - 4 | 691 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |