Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026237.7 | NP_001021408.2 | 4 - 5 | 706 | 147 | |
NM_001026238.5 | NP_001021409.2 | 8 - 9 | 1379 | 147 | ||
NM_001030057.4 | NP_001025228.2 | 10 - 11 | 1959 | 147 | ||
NM_001313465.3 | NP_001300394.1 | 15 | 2125 | 147 | ||
NM_001313466.5 | NP_001300395.1 | 16 | 2215 | 147 | ||
NM_001313491.3 | NP_001300420.1 | 1 - 2 | 358 | 147 | ||
NM_001377616.3 | NP_001364651.1 | 13 - 14 | 1804 | 147 | ||
NM_001377617.3 | NP_001364650.1 | 10 - 11 | 1528 | 147 | ||
GenBank mRNA | EF443133.1 | - | 13 - 14 | 1804 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |