Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025827.5 | NP_001020998.1 | 11 - 12 | 1941 | 79 | |
NM_001025828.6 | NP_001020999.1 | 11 - 12 | 1929 | 79 | ||
GenBank mRNA | AF195610.1 | - | 10 - 11 | 1734 | 79 | |
AF195611.1 | - | 10 - 11 | 1746 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |