Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025827.5 | NP_001020998.1 | 12 - 13 | 2319 | 86 | |
NM_001025828.6 | NP_001020999.1 | 12 - 13 | 2307 | 86 | ||
GenBank mRNA | AF195610.1 | - | 11 - 12 | 2112 | 86 | |
AF195611.1 | - | 11 - 12 | 2124 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |