Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136314.5 | NP_001129786.2 | 6 - 7 | 1076 | 97 | |
NM_060092.4 | NP_492493.2 | 6 - 7 | 1068 | 97 | ||
GenBank mRNA | AM498742.1 | - | 6 - 7 | 1068 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |