Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128955.4 | NP_001122427.1 | 8 - 9 | 1151 | 139 | |
NM_001264202.1 | NP_001251131.1 | 8 - 9 | 1151 | 217 | ||
NM_001264203.2 | NP_001251132.1 | 8 - 9 | 998 | 217 | ||
NM_060134.4 | NP_492535.1 | 7 - 8 | 989 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |