Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ile-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060147.6 | NP_492548.1 | 4 - 5 | 519 | 116 | |
GenBank mRNA | DQ384615.1 | - | 4 - 5 | 517 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |