Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sur-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060190.7 | NP_492591.1 | 1 - 2 | 314 | 140 | |
GenBank mRNA | AF174643.1 | - | 1 - 2 | 312 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |