Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hrdl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060201.6 | NP_492602.2 | 5 - 6 | 1525 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |