Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136327.4 | NP_001129799.1 | 2 - 3 | 943 | 97 | |
NM_001373584.2 | NP_001360798.1 | 5 - 6 | 1429 | 97 | ||
NM_001381160.1 | NP_001368765.1 | 2 - 3 | 934 | 97 | ||
NM_060206.7 | NP_492607.1 | 5 - 6 | 1440 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |