Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313508.2 | NP_001300437.1 | 4 - 5 | 1290 | 95 | |
NM_060234.7 | NP_492635.1 | 4 - 5 | 1290 | 95 | ||
GenBank mRNA | AF013953.1 | - | 4 - 5 | 1282 | 95 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |