Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sys-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313509.3 | NP_001300438.1 | 1 - 2 | 172 | 137 | |
NM_060238.6 | NP_492639.1 | 3 - 4 | 579 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |