Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eak-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128976.3 | NP_001122448.1 | 1 - 2 | 102 | 126 | |
NM_001128977.1 | NP_001122449.1 | 1 - 2 | 90 | 126 | ||
GenBank mRNA | DQ907009.1 | - | 1 - 2 | 90 | 126 | |
DQ907010.1 | - | 1 - 2 | 90 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |