Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128952.3 | NP_001122424.1 | 1 - 2 | 201 | 119 | |
NM_060282.6 | NP_492683.1 | 1 - 2 | 201 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |