Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060282.6 | NP_492683.1 | 8 - 9 | 1094 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |