Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mgl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313467.3 | NP_001300396.1 | 1 | 115 | 62 | |
NM_001313468.3 | NP_001300397.1 | 2 | 194 | 62 | ||
NM_060319.5 | NP_492720.3 | 1 - 2 | 187 | 62 |
Gene Symbol: maf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381197.2 | NP_001367180.1 | 1 - 2 | 194 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |