Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: blmp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001264441.2 | NP_001251370.1 | 3 - 4 | 1193 | 103 | |
NM_001264442.2 | NP_001251371.1 | 3 - 4 | 1193 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |