Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383719.1 | 2 - 3 | 185 | 71 | ||
NM_060327.7 | NP_492728.2 | 2 - 3 | 181 | 71 | ||
NR_101614.1 | 1 - 2 | 149 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |