Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: duo-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306534.2 | NP_001293463.1 | 5 - 6 | 909 | 104 | |
NM_001306535.3 | NP_001293464.1 | 2 - 3 | 444 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |