Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbtr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129070.2 | NP_001122542.1 | 2 - 3 | 370 | 106 | |
NM_001129071.3 | NP_001122543.1 | 2 - 3 | 308 | 106 | ||
GenBank mRNA | DQ904352.1 | - | 2 - 3 | 310 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |