Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tol-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025812.5 | NP_001020983.1 | 4 - 5 | 400 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |