Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: deps-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306264.3 | NP_001293193.1 | 9 - 10 | 1434 | 88 | |
NM_058341.6 | NP_490742.1 | 10 - 11 | 1573 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |