Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: deps-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306264.3 | NP_001293193.1 | 4 - 5 | 592 | 66 | |
NM_058341.6 | NP_490742.1 | 5 - 6 | 731 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |