Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: deps-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306264.3 | NP_001293193.1 | 7 - 8 | 1088 | 121 | |
NM_058341.6 | NP_490742.1 | 8 - 9 | 1227 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |