Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lpr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306337.3 | NP_001293266.1 | 1 - 2 | 77 | 87 | |
NM_001306338.4 | NP_001293267.1 | 1 - 2 | 77 | 87 | ||
NM_001306339.3 | NP_001293268.1 | 2 - 3 | 337 | 87 | ||
NM_001306340.3 | NP_001293269.1 | 2 - 3 | 268 | 87 | ||
NM_001306341.2 | NP_001293270.1 | 2 - 3 | 281 | 87 | ||
NM_058351.5 | NP_490752.2 | 2 - 3 | 337 | 87 | ||
GenBank mRNA | FJ174666.1 | - | 2 - 3 | 281 | 87 | |
FJ174667.1 | - | 2 - 3 | 268 | 87 | ||
FJ174668.1 | - | 2 - 3 | 281 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |