Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383432.1 | 7 - 8 | 857 | 66 | ||
NM_058397.7 | NP_490798.1 | 12 - 13 | 2125 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |