Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haf-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306301.3 | NP_001293230.1 | 1 - 2 | 140 | 88 | |
NM_058427.6 | NP_490828.3 | 3 - 4 | 708 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |