Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haf-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306301.3 | NP_001293230.1 | 4 - 5 | 946 | 81 | |
NM_058427.6 | NP_490828.3 | 6 - 7 | 1514 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |