Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058458.7 | NP_490859.1 | 1 - 2 | 304 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |