Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381283.2 | NP_001367687.1 | 5 - 6 | 2150 | 68 | |
GenBank mRNA | U89308.1 | - | 2 - 3 | 322 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |