Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: afd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026489.7 | NP_001021660.2 | 5 - 6 | 843 | 97 | |
NM_001026490.6 | NP_001021661.2 | 5 - 6 | 841 | 97 | ||
NM_001306400.4 | NP_001293329.1 | 5 - 6 | 845 | 97 | ||
NM_058537.5 | NP_490938.3 | 5 - 6 | 837 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |