Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306398.2 | NP_001293327.1 | 13 - 14 | 2580 | 96 | |
NM_001306399.3 | NP_001293328.1 | 13 - 14 | 2571 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |