Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zeel-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058565.4 | NP_490966.2 | 11 - 12 | 1918 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |