Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nekl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306391.3 | NP_001293320.1 | 3 - 4 | 427 | 109 | |
NM_001306392.3 | NP_001293321.1 | 2 - 3 | 313 | 109 | ||
NR_131355.1 | 3 - 4 | 427 | 109 | |||
NR_131356.1 | 2 - 3 | 316 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |