Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026642.5 | NP_001021813.1 | 8 - 9 | 2257 | 91 | |
NM_001306438.3 | NP_001293367.1 | 5 - 6 | 1021 | 91 | ||
NM_058627.7 | NP_491028.2 | 7 - 8 | 1256 | 91 | ||
NR_131370.1 | 5 - 6 | 1055 | 91 | |||
GenBank mRNA | U63557.1 | - | 7 - 8 | 1265 | 91 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |