Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cogc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026635.4 | NP_001021806.1 | 7 - 8 | 1203 | 109 | |
NM_001383914.1 | 6 - 7 | 1098 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |