Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mek-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058686.5 | NP_491087.1 | 3 - 4 | 496 | 81 | |
GenBank mRNA | U21107.1 | - | 3 - 4 | 515 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |