Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mek-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058686.5 | NP_491087.1 | 5 - 6 | 708 | 148 | |
GenBank mRNA | U21107.1 | - | 5 - 6 | 727 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |