Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mod-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058694.4 | NP_491095.3 | 8 - 9 | 1259 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |