Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rom-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058724.6 | NP_491125.4 | 4 - 5 | 527 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |