Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129055.4 | NP_001122527.1 | 11 | 1602 | 67 | |
NM_001313513.3 | NP_001300442.1 | 10 | 1087 | 67 | ||
NM_001313514.3 | NP_001300443.1 | 10 | 1543 | 67 | ||
NM_058736.5 | NP_491137.2 | 10 | 1084 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Both primers and probe map within a single exon |