Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373791.2 | NP_001360427.1 | 9 - 10 | 2090 | 85 | |
NM_058737.5 | NP_491138.2 | 9 - 10 | 2073 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |