Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sep-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058759.6 | NP_491160.1 | 4 - 5 | 1362 | 93 | |
GenBank mRNA | AF399825.1 | - | 3 - 4 | 1339 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |